missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TrueMark™ OpenArray™ Respiratory Tract Microbiota Plate
Description
- An efficient, easy-to-use OpenArray plate (112-assay format) for the characterization of key respiratory tract microbial targets through real-time PCR
- This plate contains TaqMan Microbial Assays for 31 microbial targets, plus Universal Extraction Organism Control (B. atropheus), Universal DNA Spike-In Positive Control (Xeno) and human RNase P RPPH1 control
- The format of this OpenArray plate allows for three replicates to be run in parallel per plate for all targets and two replicates for B. atropheus and RNase P
- Each OpenArray can be used for running 23 samples and 1 control sample
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | Low |
| Product Type | Respiratory Tract Microbiota Plate |
| Quantity | 1 plate |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.