missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Non-coding RNA Assay
GREENER_CHOICE

$606.00 - $2,260.00

Specifications

Content And Storage 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Product Line TaqMan
Shipping Condition Room Temperature
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
4426961
GREENER_CHOICE
View Documents Promotion Details
Applied Biosystems™
4426961
S (360 rxns), made to order
Each for $606.00
Only null left
 
4426962
View Documents Promotion Details
Applied Biosystems™
4426962
M (750 rxns), made to order
Each for $1,040.00
Only null left
 
4426963
View Documents Promotion Details
Applied Biosystems™
4426963
L (2900 rxns), made to order
Each for $2,260.00
Only null left
 
Description

Description

Specifications

Specifications

1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

TaqMan
Room Temperature
Videos
SDS
Documents

Documents