Learn More
Applied Biosystems™ TaqMan™ Non-coding RNA Assay
Supplier: Applied Biosystems™ 4426962
Chemical Identifiers
| 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
|
| TaqMan | |
| Room Temperature |
Specifications
| 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
|
| 750 reactions | |
| M (750 rxns), made to order | |
| Liquid |
| Real Time PCR (qPCR) | |
| TaqMan | |
| Room Temperature |