missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ Salmonella Enteritidis Detection Kit

Catalog No. 4457030
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4457030 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4457030 Supplier Thermo Fisher Scientific Supplier No. 4457030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage • Salmonella enteritidis 10x Assay Mix: Store at -15 °C to -25 °C
• Negative control: Store at -15 °C to -25 °C upon receipt. Store at 2 °C to 8 °C after use.
• 2x Environmental Master Mix: Store at -15 °C to -25 °C upon receipt. Store at 2 °C to 8 °C after use.
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
Product Line TaqMan
Product Type Salmonella Enteritidis Detection Kit
Quantity 1 kit
Shipping Condition Dry Ice
Target DNA
Target Organism Class S. enteriditis
For Use With (Application) Pathogen Detection
Sample Type Egg, Environmental Sample, Environmental Samples
Show More Show Less
Why are each of the individual components in the TaqMan Pathogen Detections Kits color-coded?

Each individual component in the TaqMan Pathogen Detection Kits is color-coded in order to best distinguish between the 2X Environmental Master Mix, the negative control and the 10X Target Assay Mix (TAM). Yellow coded vials are 2X Environmental Master Mix, the white coded vial is the negative control solution, and the 10X Target Assay Mix (TAM) which is associated with a specific organism is a gray-labeled vial.

For Research Use Only. Not for use in diagnostic procedures.