missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Fast Reagent Starter Kit

Catalog No. 4352407
Encompass
Change view
Click to view available options
Quantity:
125 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4352407 125 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4352407 Supplier Applied Biosystems™ Supplier No. 4352407
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to detect and quantify Eukaryotic 18S rRNA sequence in cDNA samples using the TaqMan 5' nuclease assay, in fast thermal cycling mode.

The Eukaryotic 18S rRNA TaqMan™ Assay consists of a 20X mix of two unlabeled PCR primers and a FAM™ dye labeled 18S TaqMan™ MGB probe. Used With: 7900HT Fast Real-Time PCR System with Fast 96-well Block Module, 7900HT Fast Real-Time Service Upgrade, 7900HT System Fast 96-well Block Upgrade Kit.

Order Info

Dry Ice

Specifications

Content And Storage This kit contains TaqMan™ Fast Real-Time PCR Universal Master Mix (2X), no AmpErase™ UNG (1 tube) , Eukaryotic 18S rRNA TaqMan™ Assay (20X mix of two unlabeled PCR primers and a FAM™ dye-labeled 18S TaqMan™ MGB probe) (Assay ID Hs99999901_s1; Part Number 4331182), and human Raji cDNA.

Store the PCR mix at 2° to 8°C
Store the rest of the kit contents at -15° to -25°C.
Detection Method Primer-probe
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, AB 7900HT Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line TaqMan
Product Type PCR Reagents
Quantity 125 reactions
Shipping Condition Dry Ice
Sufficient For 125 Reactions
Target 18S rRNA
Target Specificity Target-Specific
For Use With (Application) Gene Expression
Hot Start No
No. of Reactions 125 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.