missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan Fast Advanced Master Mix Gene Expression Bundle

Catalog No. A47374
Encompass
Change view
Click to view available options
Includes:
Master Mix
Master Mix, MicroAmp Fast Plates, and MicroAmp Seals
Master Mix, MicroAmp Plates, and MicroAmp Seals
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
A47374 Master Mix
A47377 Master Mix, MicroAmp Plates, and MicroAmp Seals
A47376 Master Mix, MicroAmp Fast Plates, and MicroAmp Seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A47374 Supplier Applied Biosystems™ Supplier No. A47374
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

TaqMan Fast Advanced Master Mix delivers better sensitivity, greater specificity, and faster results

Applied Biosystems has affordable qPCR tools that fit every budget and level of experience. Makes getting the best results from your data easy while saving money.

Specifications

Content And Storage • Master Mix, 5 mL
Description Gene Expression Starter Kit
For Use With (Application) Real Time PCR (qPCR)
Includes Master Mix