missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents
GREENER_CHOICE

Catalog No. 4308321
Encompass
Change view
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
4308321 1000 Reactions
4308323 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4308321 Supplier Applied Biosystems™ Supplier No. 4308321
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe.

  • Applied Biosystems™ TaqMan™ Exogenous Internal Positive Control Reagents contain a pre-optimized internal positive control (IPC) with pre-designed primers and TaqMan™ probe
  • The IPC can be spiked into samples to distinguish true target negatives from PCR inhibition
  • This kit allows users to amplify a low-copy target DNA in the same tube with the IPC
  • Although the target and IPC DNAs may differ in initial copy number, the concentration of the IPC primers in the PCR reaction is limiting so that the amplification efficiency of the target reaction is not compromised
  • Pre-designed primers and TaqMan™ probe eliminate assay design
  • Rapid assay development guidelines can minimize optimization time Separate components provide flexibility in assay set-up
  • Reagents may be stored at 2°C to 4°C if they are to be consumed within one month
  • Otherwise, store at -20°C to -25°C

Note: This is not a quantitative kit and should not be used for normalization Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence is present, and a negative call for the target sequence and negative call for the IPC suggest PCR inhibition.

Order Info

Note: This is not a quantitative kit and should not be used for normalization. Instead, this kit is designed to distinguish types of negative results: a negative call for the target sequence and a positive call for the IPC indicate that no target sequence

Specifications

Content And Storage Contains 5 of the following:
• 1 ml 10X Exo IPC Mix
• 120 μl 10X Exo IPC Block
• 200 μl 50X Exo IPC DNA

Store at -20°C to -25°C. Reagents may be stored at 2–4°C if they are to be consumed within one month.
Format Tube(s)
GC-Rich PCR Performance Low
PCR Method qPCR
Reaction Speed Standard
Internal Probe Modification TAMRA™ Quencher (3'), VIC™ (5')
Product Line TaqMan
Product Type Exogenous Internal Positive Control Reagent
Quantity 1,000 reactions
Sufficient For 1000 Reactions
No. of Reactions 1000 Reactions
Show More Show Less
Can I order TaqMan Exogenous Internal Positive Control Reagents (Cat. Nos. 4308321, 4308323) with a different quencher?

Yes, you can order TaqMan Exogenous Internal Positive Control Reagents with QSY through Specialty Oligos with no need for redesign. You may reach out to our Specialty Oligos team at Specialty_Oligos@thermofisher.com.
You can also order them with MGB, but this would require redesign, and they would need to be made with bioinformatics through the Assay Design Hub, if possible.

For Research Use Only. Not for use in diagnostic procedures.