missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Copy Number Reference Assay, human, TERT

Catalog No. 4403315
Change view
Click to view available options
No. of Reactions:
3000 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
4403315 3000 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4403315 Supplier Applied Biosystems™ Supplier No. 4403315
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

TaqMan™ Copy Number Reference Assay, human, TERT

Human TaqMan™ Copy Number Reference Assays are run with human TaqMan™ Copy Number Assays in a duplex real-time PCR reaction to detect and measure copy number variations (CNVs) and smaller regions in the human genome. Two options for genes that can be used as endogenous reference genes in humans are offered: RNase P and TERT. TaqMan™ Copy Number Reference Assays are designed to unique genomic sequences in the reference genome assembly (Build 37/hg19) and are required for relative quantitation of copy number targets.

TaqMan™ Copy Number Assays quantitate the gene of interest and normalize to an endogenous reference gene known to be present in two copies in a diploid genome. Please note that TaqMan™ Copy Number Reference Assays are species specific.

TERT: The Alternative Reference Gene Option
TaqMan™ Copy Number Reference Assays have a VIC™ dye–labeled TAMRA™ probe and reference sequence–specific forward and reverse primers. The assays are not primer-limited.

TaqMan™ Copy Number Reference Assay RNase P is recommended as the standard reference assay for copy number analysis. This assay detects the Ribonuclease P RNA component H1 (H1RNA) gene (RPPH1) on chromosome 14, cytoband 14q11.2. The assay location is chr.14:20811565 on NCBI build 37. It has an 87 bp amplicon that maps within the single exon RPPH1 gene.

TaqMan™ Copy Number Reference Assay TERT is an alternative reference assay; it is recommended in the event that the RNase P assay functions poorly with a sample because of chromosomal aberrations or other issues. This assay targets the telomerase reverse transcriptase (TERT) gene located on chromosome 5, cytoband 5p15.33. The assay location is chr.5:1253373 on NCBI build 37. It has an 88 bp amplicon that maps within exon 16 of the TERT gene.

Simplest Copy Number Analysis Workflow
TaqMan™ Copy Number Assays have the simplest workflow of all currently available copy number analysis methods. The test assay (FAM™ dye–labeled), the reference assay (VIC™ dye–labeled), your sample DNA, and TaqMan™ Master Mix are combined and then run on an Applied Biosystems™ real-time PCR system using the standard TaqMan™ Copy Number Assay protocol. On average, setup to primary analysis takes only 3–4 hr (including an approximately 2 hour PCR run).

Powerful Data analysis Software
CopyCaller™ Software was developed specifically for TaqMan™ Copy Number Assay data analysis. This free, easy-to-use software utilizes a graphical interface that quickly calculates the possible copy numbers for a set of samples in a run. It also gives a confidence value for each copy number call, and has outlier functionality.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Specifications

3'Primer Modification None
5'Primer Modification None
Concentration 20X
Content And Storage Control Primer-Probe Set(s)
Detection Method Primer-probe
Form Frozen
Format Tube
GC-Rich PCR Performance Low
PCR Method qPCR
Reaction Speed Standard
Gene Symbol TERT
Internal Probe Modification TAMRA™ Quencher (3'), VIC™ (5')
Label or Dye VIC
Product Line TaqMan
Product Type Configurable Assay
Purification Method HPLC
Purity or Quality Grade RNase-Free, DNase-Free
Quantity 3,000 reactions
Shipping Condition Dry Ice
Species Human
Sufficient For 3000 Reactions
Target Tert (Telomerase Reverse Transcriptase)
No. of Reactions 3000 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.