Learn More
Invitrogen™ TaqMan™ Cells-to-CT™ HepatoExpress™ Kit

Description
The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a hepatocyte gene expression kit designed for RNA extraction-free RT-qPCR. It enables fast, direct lysis of primary hepatocytes, eliminating the need for RNA purification while reducing sample preparation time to ~10 minutes.
This streamlined cell-to-CT workflow allows researchers to perform high-sensitivity gene expression analysis directly from cells, without columns, beads, or centrifugation. The result is reliable, reproducible RT-qPCR data with significantly reduced hands-on time.
Optimized specifically for primary hepatocyte models—including ECM-overlay cultures—the kit supports high-throughput, automation-compatible workflows for drug metabolism, toxicity studies, and mechanistic research.
Key Benefits
Fast, Extraction-Free Gene Expression Workflow
- Perform RT-qPCR without RNA isolation or purification
- Reduce RNA preparation time from hours to ~10 minutes
- Direct lysis workflow minimizes sample handling and variability
High-Quality RT-qPCR Performance
- Sensitivity and reproducibility comparable to purified RNA workflows
- Preserves RNA integrity during lysis
- Compatible with multiplex qPCR and low-input samples
Optimized for Primary Hepatocytes
- Efficient lysis of hepatocytes, including ECM-overlay cultures
- Consistent performance across hepatocyte species and models
- Designed for challenging in vitro liver systems
Automation-Ready and Scalable
- Compatible with 96-well plate formats and robotic liquid handling systems
- An ideal choice for high-throughput screening and large-scale studies
- Room-temperature workflow simplifies lab integration
Sustainable and Simplified Workflow
- Significantly reduces plastic consumables associated with RNA purification
- REACH-compliant reagents support environmentally responsible lab practices
Designed for Hepatocyte Gene Expression Challenges
Primary hepatocytes are widely used for drug metabolism, CYP450 induction, and toxicity studies, but efficient RNA extraction from these cells—especially in ECM-overlay cultures—can be difficult and time-consuming.
The HepatoExpress Kit addresses these challenges with a direct lysis chemistry that:
- Effectively penetrates extracellular matrix (ECM) overlays
- Protects RNA during lysis without purification
- Maintains compatibility with reverse transcription and qPCR
This enables accurate gene expression analysis without the variability introduced by traditional RNA extraction workflows.
Applications
The HepatoExpress Kit is an ideal choice for:
- Hepatocyte gene expression analysis
- CYP450 induction assays
- Drug metabolism and ADME studies
- Toxicology and safety screening
- Target verification and early-stage compound screening
- Mechanistic studies in liver biology
Scalable from Discovery to High-Throughput Screening
Whether performing small-scale mechanistic studies or large-scale screening, the kit supports flexible experimental design. Its automation-friendly format enables seamless integration into high-throughput workflows, improving reproducibility and throughput.
Workflow: From Cells to qPCR in ~75 Minutes
The simple three-step workflow enables rapid gene expression analysis:
- Cell lysis (~10 minutes, room temperature)
- Simultaneous cell lysis, RNA preservation, and genomic DNA removal
- Reverse transcription
- cDNA synthesis using SuperScript™ IV VILO™ Master Mix
- qPCR detection
- Sensitive, multiplex-capable amplification using TaqMan™ Gene Expression Assays
Compatible with 700,000+ TaqMan™ Gene Expression Assays, enabling broad pathway and target coverage.
Why Choose an RNA Extraction-Free qPCR Workflow?
Compared to traditional column-based RNA isolation methods, the HepatoExpress Kit:
- Reduces workflow time from hours to minutes
- Minimizes sample loss and variability
- Reduces multiple handling steps that can degrade RNA
- Improves consistency across replicates and experiments
The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a fast, reliable, and scalable solution for primary hepatocyte gene expression analysis. By enabling direct, RNA extraction-free RT-qPCR, it simplifies workflows while maintaining high data quality—making it excellent for modern drug discovery and in vitro liver research.
Specifications
Specifications
| Content And Storage | HepatoExpress Lysis Solution (Room Temperature), HepatoExpress LysisGuard (-20°C), Express ezDNase (-20°C). For long-term storage, store HepatoExpress Lysis Buffer at 4°C. |
| Detection Method | TaqMan |
| Form | Liquid |
| Format | Kit |
| GC-Rich PCR Performance | High |
| PCR Method | 2-step RT-qPCR |
| Reaction Speed | 10 min. |
| High-throughput Compatibility | Yes |
| Label or Dye | ROX |
| Product Type | Direct Cell Lysis to RT-qPCR Kit |
| Show More |
Frequently Asked Questions (FAQs)
The TaqMan Cells-to-CT HepatoExpress Kit has not been tested with 3D cultures.
The TaqMan Cells-to-CT HepatoExpress Kit has not been tested with tissue cells in 384-well plates.
No, ezDNase contained in the TaqMan Cells-to-CT HepatoExpress Kit is a double-strand specific DNase and will not degrade cDNA produced in the RT reaction. It is also heat-labile and will be inactivated during the heat kill step of the RT reaction.
With the TaqMan Cells-to-CT HepatoExpress Kit, cells can be washed using warm or cold wash buffer according to user preference. In our evaluation, we found that using warm or chilled wash buffer yielded equivalent results.
Yes, the TaqMan Cells-to-CT HepatoExpress Kit allows for direct RT-qPCR amplification from cell lysates without purification of RNA.
For Research Use Only. Not for use in diagnostic procedures.