missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Respiratory Tract Microbiota Comprehensive Card

Catalog No. A41238
Encompass
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41238 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41238 Supplier Applied Biosystems™ Supplier No. A41238

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TaqMan™ Array Respiratory Tract Microbiota Comprehensive Card is an efficient, easy-to-use TaqMan™ Array Card (48-assay format) for the characterization of key respiratory tract microbial targets through real-time PCR.

  • An efficient, easy-to-use TaqMan Array Card (48-assay format) for the characterization of key respiratory tract microbial targets through real-time PCR
  • This card contains TaqMan Microbial Assays for 42 microbial targets, plus Universal Extraction Organism Control (B. atropheus), Universal DNA Spike-In Positive Control (Xeno), 18s control and human RNase P RPPH1 control
  • Each TaqMan Array Card can be used for running 7 samples and 1 control sample

Key Features of TaqMan Array Cards:

  • Pre-formatted and inventoried - each card contains a select group of TaqMan Microbial Assays pre-configured in a micro fluidic array, ready to ship
  • Convenient and easy to use - get the performance users have come to expect from TaqMan Microbial Assays without the need for expensive robotics
  • Reproducible results - get consistent, reliable data across samples, studies, and labs, the same high data quality from card-to-card and lot-to-lot, even with different operators

Specifications

Content And Storage Store refrigerated at 4°C.
Product Line TaqMan
Quantity 1 Plate
Shipping Condition Approved for shipment at ambient temperature
For Use With (Application) Pathogen Detection, Virus Detection

For Research Use Only. Not for use in diagnostic procedures.