missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Mouse Stem Cell Pluripotency Panel

Catalog No. 4385363
Change view
Click to view available options
Quantity:
4 cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4385363 4 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4385363 Supplier Applied Biosystems™ Supplier No. 4385363

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Form Dried Down
Format 384-well microfluidic card
Reaction Speed Standard
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Research Category Cell Cycle & Proliferation, Biomarker-Related Pathway(s), Stem Cell Research, Reproduction
Species Mouse
Target Mouse orthologs of well-defined human markers for stem cell pluripotency
For Use With (Application) Stem Cell Analysis
No. of Reactions 1 x 2 μL Reactions
Show More Show Less
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.