missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array Human Inflammation Panel

Catalog No. 4378707
Change view
Click to view available options
Quantity:
4 cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4378707 4 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4378707 Supplier Applied Biosystems™ Supplier No. 4378707
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems™ TaqMan™ Human Inflammation Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery.

  • An efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of inflammation genes important in drug discovery
  • Designed with 96 TaqMan™ Gene Expression Assays arrayed in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and is sold in packs of 4
  • TaqMan™ Gene Signature Arrays for human inflammation contain over 90 genes that are, or have been, studied as targets for a range of inflammatory diseases
  • The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics
  • Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators

Specifications

Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Form Dried Down
Format 384-well microfluidic card
Reaction Speed Standard
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Research Category Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Inflammation
Species Human
Target Over 90 genes that are targets for a range of inflammatory diseases
For Use With (Application) Gene Expression
No. of Reactions 1 x 2 μL Reactions
Show More Show Less
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.