Learn More
Applied Biosystems™ TaqMan™ Array Human DNA Methylation & Transcriptional Repression
Description
- Extensive collections - choose from over 130 predesigned TaqMan™ Gene Signature Arrays and Plates, or customize your own plate, selecting from any of our 1.3 million inventoried assays across 24 species
- Affordable - minimum order of only one gene signature plate or six custom plates
- Convenient - dried TaqMan probe and primer sets supplied in 96-well plates; just add master mix and sample, and then run
- Quick results - Fast format typically provides results in under an hour
Specifications
Specifications
| Content And Storage | Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C. |
| Form | Dried Down |
| Format | 96-well plate |
| Reaction Speed | Standard |
| Internal Probe Modification | MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5') |
| Label or Dye | FAM |
| Product Line | TaqMan |
| Purification Method | Standard |
| Quantity | 1 standard plate |
| Research Category | Signal Transduction, Gene Regulation, Epigenetics, Oncology |
| Show More |
Frequently Asked Questions (FAQs)
The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.
We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.
Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch
Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.
For Research Use Only. Not for use in diagnostic procedures.