Learn More
TaqMan™ Array, Format 96b
Catalog No. 4342261
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
4342261 1 each
1 options
Catalog No. 4342261 Supplier Thermo Fisher Scientific Supplier No. 4342261
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors.
The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.
Specifications
Content And Storage Store in refrigerator (2°C to 8°C)
Product Line TaqMan
Quantity 1 each
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.
For Research Use Only. Not for use in diagnostic procedures.