missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Array, Format 96a
Description
The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). |
| Format | 96A |
| Product Line | TaqMan |
| Product Type | Custom Gene Expression TaqMan™ Array Cards |
| Quantity | 1 Ea. |
Frequently Asked Questions (FAQs)
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.
For Research Use Only. Not for use in diagnostic procedures.