missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Format 48

Catalog No. 4342253
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4342253 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4342253 Supplier Applied Biosystems™ Supplier No. 4342253
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors.

The Custom TaqMan® Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors. Select target assays from thousands of inventoried TaqMan® Gene Expression Assays.

Specifications

Content And Storage Store in refrigerator (2–8°C).
Detection Method Primer-probe
Form Liquid
Format 384-Well microfluidic card, Format 48
PCR Method qPCR
Product Line TaqMan
Product Type Custom Taqman Array
Quantity 1 each
No. of Reactions 384 Reactions
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.