missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Array, Format 24

Catalog No. 4342249
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4342249 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4342249 Supplier Applied Biosystems™ Supplier No. 4342249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Custom TaqMan™ Array Card is a 384-well microfluidic card designed to perform 384 simultaneous real-time PCR reactions without using liquid-handling robots or multichannel pipettors.

Select target assays from thousands of inventoried TaqMan™ Gene Expression Assays.

Specifications

Content And Storage Store in refrigerator (2–8°C).
Detection Method Primer-probe
Form Liquid
Format 384-Well microfluidic card, Format 24
PCR Method qPCR
Product Line TaqMan
Product Type TaqMan Array Cards
Quantity Each
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.