missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Arbovirus Triplex Kit (ZIKV/DENV/CHIKV), 0.2 mL

A simple solution for dengue, chikungunya, and Zika virus RNA detection: just add RNA.

Supplier:  Applied Biosystems™ A31746

Catalog No. A31746


Only null left
Explore available promotions

Description

Description

The TaqMan Arbovirus Triplex Kit enables the simultaneous detection and differentiation of RNA from dengue virus (DENV), chikungunya virus (CHIKV), and Zika virus (ZIKV) in urine and serum. It is a lyophilized, multiplex, one-step real-time PCR Applied Biosystems TaqMan assay for the molecular detection of dengue, chikungunya, and Zika RNA. The kit is optimized for use with the Applied Biosystems QuantStudio 5 Real-Time PCR System, but is also suitable for the 7500 and other QuantStudio systems. It includes a primer-probe set for human PPIA (cyclophilin A) as an endogenous extraction, RT-PCR and PCR amplification control. The kit is compatible with RNA extracted from human urine or serum using the MagMAX Pathogen RNA/DNA Kit. The Zika virus sequence used to develop these primers and probes is from a 2014 research paper by Pyke et al (1).

Features of the kit:
• Lyophilized with one-step RT-PCR master mix; simply add RNA
• Simple workflow for detection of Zika, Dengue, and Chikungunya virus RNA from urine or serum
• Easy to use and environmentally friendly. No cold chain management required; shipped and stored at room temperature.

Contents:
• Twelve 8-well strips tubes for 96 total reactions per kit
• 0.2-mL tube format, compatible with 96-well block
• Primer-probe detection with 4 probe reporter dyes:
   --FAM-Zika
   --VIC-Dengue
   --ABY-Chikungunya
   --JUN-PPIA cyclophilin

1. Pyke AT, Daly MT, Cameron JN, Moore PR, Taylor CT, Hewitson GR, Humphreys JL, Gair R. Imported zika virus infection from the cook islands into australia, 2014. PLoS Curr. 2014 Jun 2;6.

Order Info

Shipping Conditions: Ambient Temperature

Specifications

Specifications

No cold-chain management required, ship and store at room temperature.
Primer-probe
TaqMan
Pathogen Detection
Multiplex standard strip (0.2mL, 12 x 8-well strip tubes)
Any qPCR insturment
0.2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions