missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqCheck™ SARS-CoV-2 Control

Catalog No. 956127
Encompass
Change view
Click to view available options
Quantity:
3 x 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
956127 3 x 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 956127 Supplier Applied Biosystems™ Supplier No. LS956127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems™ TaqCheck™ SARS-CoV-2 Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 S and N gene assays included in the TaqCheck™ SARS-CoV-2 Fast PCR Assay.

The Applied Biosystems TaqCheck SARS-CoV-2 Control is a synthetic RNA positive control that contains target sequences for the SARS-CoV-2 S and N gene assays included in the TaqCheck SARS-CoV-2 Fast PCR Assay.

Specifications

PCR Method 1-step RT-qPCR
Reaction Speed Fast
For Use With (Equipment) QuantStudio™ 5 Real‐Time PCR Instrument, 384-well
Product Line TaqCheck
Quantity 3 x 10 μL
Shipping Condition Dry Ice
Species Human
Target Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
For Use With (Application) Real Time PCR (qPCR)

For Research Use Only. Not for use in diagnostic procedures.