Learn More
MP Biomedicals™ Taq CORE Kit 25
Supplied with standard reaction buffers
Supplier: MP Biomedicals™ 11EPTQK259
Includes: Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix
Specifications
| Taq DNA Polymerase 5U/μL, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 25mM dNTP Mix. Store at -20°C | |
| 10 x 250U | |
| Standard PCR |