missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPSO, for biochemistry

CAS: 68399-81-5 | C7H17NO7S | 259.273 g/mol

$168.76 - $168.76

Chemical Identifiers

CAS 68399-81-5
Molecular Formula C7H17NO7S
Molecular Weight (g/mol) 259.273
MDL Number MFCD00038352
InChI Key RZQXOGQSPBYUKH-UHFFFAOYSA-N
Synonym 3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid
PubChem CID 109334
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC327811000
SDS View Documents
Thermo Scientific Chemicals
327811000
100 g Glass bottle
Each for $168.76
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

68399-81-5
259.273
RZQXOGQSPBYUKH-UHFFFAOYSA-N
109334
C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO
C7H17NO7S
MFCD00038352
3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid

Specifications

(0.1 M aq. soln.)
220.0°C to 221.0°C
White to Yellow
Powder
99+%
C7H17NO7S
MFCD00038352
3-(N-Tris-(hydroxymethyl)methylamino)-2-hydroxypropanesulfonic acid
RZQXOGQSPBYUKH-UHFFFAOYSA-N
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]-2-hydroxypropane-1-sulfonic acid
109334
≥99% (Acid)
TAPSO
(0.1M aq. soln.)
68399-81-5
4.3 to 5.3 (1% aq. soln. at 25°C)
100 g
Authentic
(HOCH2)3CNHCH2CH(OH)CH2SO3H
Glass bottle
Solubility in water: ca 129.6g/L (20°C).
C(C(CS(=O)(=O)O)O)NC(CO)(CO)CO
259.273
259.3
Biochemical
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only