missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ TAPS, ULTROL™ Grade, Calbiochem™,

Catalog No. 3946751KG
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3946751KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3946751KG Supplier MilliporeSigma™ Supplier No. 3946751KG

A zwitterionic buffer useful in the pH range of 7.7 to 9.1

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym 3-{[tris(Hydroxymethyl)methyl]amino}propanesulfonic Acid
PubChem CID 121591
IUPAC Name 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Quantity 1 kg
Assay Percent Range >98%
Solubility Information Easily soluble in cold water
Formula Weight 243.2
Grade by titration
Chemical Name or Material zwitterionic buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).