missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TAPS, 99+%, for biochemistry
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Absorbance | (0.1 M aq. soln.) |
| Absorbance Conditions | (0.1M aq. soln.) |
| Melting Point | 240.0°C |
| Color | White |
| pH | 5 to 6.5 (1% aq. soln. at 25°C) |
| Physical Form | Powder |
| Quantity | 25 g |
| Flash Point | >110°C |
| Assay Percent Range | 99+% |
| Infrared Spectrum | Authentic |
| Show More |
RUO – Research Use Only