missing translation for 'onlineSavingsMsg'
Learn More

Tanshinone IIA 97.0+%, TCI America™

$210.75 - $717.67

Chemical Identifiers

CAS 568-72-9
Molecular Formula C19H18O3
Molecular Weight (g/mol) 294.35
MDL Number MFCD00238692
InChI Key HYXITZLLTYIPOF-UHFFFAOYSA-N
Synonym tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
PubChem CID 164676
IUPAC Name 1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
SMILES CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
T298710MG
View Documents Promotion Details
N/A 10 mg
Each for $210.75
Only null left
 
T298750MG
View Documents Promotion Details
N/A 50 mg
Each for $717.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

568-72-9
294.35
HYXITZLLTYIPOF-UHFFFAOYSA-N
164676
CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
C19H18O3
MFCD00238692
tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione

Specifications

568-72-9
C19H18O3
tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
294.35
294.35
Crystalline Powder
198°C
MFCD00238692
HYXITZLLTYIPOF-UHFFFAOYSA-N
1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
164676
≥97.0% (HPLC)
Tanshinone IIA
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SF8282695

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents