missing translation for 'onlineSavingsMsg'
Learn More

Tanshinone IIA 97.0+%, TCI America™

Catalog No. T298710MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T298710MG 10 mg
T298750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T298710MG Supplier TCI America Supplier No. T298710MG

Chemical Identifiers

CAS 568-72-9
Molecular Formula C19H18O3
Molecular Weight (g/mol) 294.35
MDL Number MFCD00238692
InChI Key HYXITZLLTYIPOF-UHFFFAOYSA-N
Synonym tanshinone iia, tanshinone ii, tanshinone b, dan shen ketone, tanshinon ii, 1,6,6-trimethyl-6,7,8,9-tetrahydrophenanthro 1,2-b furan-10,11-dione, unii-4gpc9fqg6l, 4gpc9fqg6l, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g benzofuran-10,11-dione, 1,6,6-trimethyl-8,9-dihydro-7h-naphtho 1,2-g 1 benzofuran-10,11-dione
PubChem CID 164676
IUPAC Name 1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
SMILES CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21

Specifications

Melting Point 198°C
Quantity 10 mg
Formula Weight 294.35
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tanshinone IIA
RTECSNumber SF8282695
TSCA No
Recommended Storage Refrigerator