accessibility menu, dialog, popup

UserName

CAS RN 568-72-9

IUPAC Name: 1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
Synonyms: tanshinone iia
Molecular Weight (g/mol): 294.35
Molecular Formula: C19H18O3
InChi Key: HYXITZLLTYIPOF-UHFFFAOYSA-N
SMILES: CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results