missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tamoxifen

Catalog No. 361063892
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361063892 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361063892 Supplier Tocris Bioscience™ Supplier No. 6342/100

Estrogen receptor partial agonist/antagonist

Tamoxifen is a selective estrogen receptor modulator (SERM). Exhibits antiestrogenic effects in mammary tissue and estrogen-like effects in bone. Anticancer. Tamoxifen citrate (Cat.No. 0999) is also available.

Specifications

Chemical Name or Material 2-[4-[(1Z)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine
CAS 10540-29-1
Quantity 100 mg
Target Estrogen and Related Receptor Modulators
Molecular Formula C26H29NO
Purity 0.98