missing translation for 'onlineSavingsMsg'
Learn More

TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™

Catalog No. bp133220
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP133220 20 L Poly CUBE
BP1332500 500 mL Poly Bottle
BP13321 1 L Nalgene Poly Bottle
BP13324 4 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP133220 Supplier Fisher BioReagents Supplier No. FLBP1332 20
Only null left

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 50X TAE Solution - simply dilute as needed
  • 50X solution contains 2M tris-acetate and 0.050M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 50X TAE
  • Solution with 49 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 50X Solution
CAS 77-86-1, 60-00-4, 6850-28-8
Physical Form Liquid
Quantity 20 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Poly CUBE
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (24%), Acetic Acid (5.0%), and EDTA (<2%).
Show More Show Less