Learn More
Sartorius Tacta™ Mechanical Pipette, Trade-In
Upgrade your pipetting with unbeatable prices. For a limited time, you can get up to 35% savings on Tacta Mechanical Pipettes.
Supplier:  Sartorius LH729120PR
 View more versions of this product
Catalog No. 14577801PR
Only null left
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.
Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.
Specifications
Chemical Identifiers
Yes
Reusable
Polypropylene
No
Pipette
Autoclavable
Ergonomic
Tacta
Non-sterile
Specifications
Optifit 10 μL, SafetySpace Filter Tips 10 μL
Autoclavable
Reusable
Polypropylene
Tacta
Non-sterile
0.5 to 10 μL
Yes
Gray
Ergonomic
8
No
Pipette
Product Suggestions
Product Suggestions