Learn More
Sartorius Tacta™ Mechanical Pipette, Trade-In
Catalog No. p-6895030
Change view
Click to view available options
Number of Channels:
1
12
8
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels Volume (Metric) Color
14577793PR 1 0.1 to 3 μL Gray
14577794PR 1 0.5 to 10 μL Gray
14577795PR 1 2 to 20 μL Yellow
14577796PR 1 10 to 100 μL Yellow
14577797PR 1 20 to 200 μL Yellow
14577798PR 1 100 to 1000 μL Blue
14577799PR 1 500 to 5000 μL Green
14577800PR 1 1000 to 10,000 μL Red
14577801PR 8 0.5 to 10 μL Gray
14577802PR 8 5 to 100 μL Yellow
14577803PR 8 30 to 300 μL Orange
14577804PR 12 0.5 to 10 μL Gray
14577805PR 12 5 to 100 μL Yellow
14577806PR 12 30 to 300 μL Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 14577793PR Supplier Sartorius Supplier No. LH729010PR
Only null left
Upgrade your pipetting with unbeatable prices. For a limited time, you can get up to 35% savings on Tacta Mechanical Pipettes.
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.
Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.
Specifications
Compatible Tips Optifit 10 μL, SafetySpace Filter Tips 10 μL
Adjustable Tip Spacing Yes
Autoclavable Autoclavable
Color Gray
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line Tacta
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.1 to 3 μL
Show More Show Less