missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences T5 Exonuclease

Catalog No. S000756
Encompass
Change view
Click to view available options
Quantity:
1000 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000756 1000 U
S000750 5000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S000756 Supplier G-Biosciences Supplier No. 7862195

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T5 Exonuclease is a highly efficient 5'-to-3' exonuclease for either ssDNA or dsDNA. It also has ssDNA specific endonuclease activity in the presence of magnesium ions.

The mode of action of T5 Exonuclease in vivo may be analogous to that of the 5-to-3 exonuclease activity of E. coli DNA Polymerase I. Applications include plasmid mutagenesis methods, oligonucleotide site-directed mutagenesis, generation of plasmid-sequencing templates, generation of 3'-overhang for improved cloning procedures.

Specifications

Packaging Vial
Enzyme Exonuclease
Sensitive to Heat Inactivation No
Quantity 1000 U
Product Type Exonuclease
Research Category Traditional Cloning
Product Line Invitrogen
Form Liquid