missing translation for 'onlineSavingsMsg'
Learn More

G-Biosciences T4 UvsX DNA Recombinase

Catalog No. S03128
Encompass
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03128 100 μg
S03127 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03128 Supplier G-Biosciences Supplier No. 7861989

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.

  • T4 UvsX DNA Recombinase forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule.
  • This process is aided by ssDNA binding proteins and recombination mediators.
  • The filament is then elongated by DNA polymerase with the newly synthesized strand displacing the old strand.
  • The newly synthesized duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA

Specifications

Sensitive to Heat Inactivation Yes
Quantity 100 μg
Product Type DNA Recombinase
Research Category PCR, Next-generation Sequencing, RNA Research, Genome Editing, Traditional Cloning
Form Liquid