missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ T 5224
Description
T 5224 is a selective inhibitor of c-Fos/activator protein (AP)-1. It inhibits the DNA-binding and transactivation activities of AP-1, without affecting the activity of ATF2, C/EBPα, NF-κB/p65, MyoD, and Sp1 in vitro. In a mouse model, T 5224 prevents type II collagen-induced arthritis by reducing inflammatory cytokines (including interleukin 1β) and matrix-degrading MMPs.
Specifications
Specifications
| Chemical Name or Material | 5-[4-(Cyclopentyloxy)-2-hydroxybenzoyl]-2-[(2,3-dihydro-3-oxo-1,2-benzisoxazol-6-yl)methoxy]benzenepropanoic acid |
| CAS | 530141-72-1 |
| Quantity | 50 mg |
| Target | AP-1 Inhibitors |
| Molecular Formula | C29H27NO8 |
| Purity | 0.98 |