Learn More
Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments
Catalog No. p-4525939
Change view
Click to view available options
Volume (Metric):
250 μL
500 μL
2.5 mL
For Use With (Equipment):
AS1000, AS3000 Autosampler
AS3000, AS3500 Autosampler
LCQ HPLC
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
365ILT21 250 μL LCQ HPLC
365JLT41 500 μL LCQ HPLC
365ILT91 250 μL AS1000, AS3000 Autosampler
365JLT61 500 μL AS1000, AS3000 Autosampler
365LLT81 2.5 mL AS3000, AS3500 Autosampler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 365ILT21 Supplier Thermo Scientific™ Supplier No. 365ILT21
Only null left
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Specifications
Needle Gauge 22 G
Length (English) Needle 2 in. Removable Needle
Product Type HPLC Autosampler Syringe
Disposable Reusable
Volume (Metric) 250 μL
For Use With (Equipment) LCQ HPLC
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.