Learn More
Synthware™ Three Neck Flask, Angled
Three Neck Flask with standard taper outer joints on both center and angled side necks.
Supplier:  Synthware™ F439250
 View more versions of this product
Catalog No. 31501253
Only null left
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness.
Specifications
Specifications
Three Neck Round Bottom Flasks, Angled
Borosilicate Glass (3.3)
Center: 19/22, Side: 19/22
Reusable
Beaded
Round
250 mL
Three-neck Flask
Autoclavable
No
Non-sterile
Product Suggestions
Product Suggestions