Learn More
Synthware™ Single Neck Round Bottom Flasks, Medium Length Joint
Hand-blown from tubing to ensure uniform wall thickness
Supplier: Synthware™ F309200
Description
SYNTHWARE round bottom boiling flasks with full length standard taper outer joint. All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness.Specifications
| Clear | |
| 200 mL | |
| Organic Synthesis |
| Single neck, round bottom flask | |
| Borosilicate Glass (3.3) | |
| 19/22 |