Learn More
Synthware™ Round Bottom Flasks with Spherical Joint
Short neck flask with spherical socket joint
$81.07 - $276.43
Specifications
| Description | Round bottom flask with spherical joint |
|---|---|
| Standard Taper Joint | 35/20 |
Description
- Heavy wall design
- All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
| Round bottom flask with spherical joint | |
| 35/20 |