Learn More
Synthware™ Round Bottom Flasks with Spherical Joint
Short neck flask with spherical socket joint
Supplier:  Synthware™ F527000
 View more versions of this product
Catalog No. 31501140
Only null left
Explore available promotions
Description
Description
  • Heavy wall design
  • All sizes hand blown from tubing to ensure uniform wall thickness
Specifications
Specifications
Round bottom flask with spherical joint
1000 mL
35/20
Product Suggestions
Product Suggestions