Learn More
Synthware™ Round Bottom Flasks with Hooks
Short neck with standard taper outer joint
$57.87 - $68.30
| Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Standard Taper Joint | Price | Quantity | ||||
|
31501150
|
Synthware™
F331405 |
5 mL | 14/20 |
Each for $57.87
|
|
||||
|
31501151
|
Synthware™
F331410 |
10 mL | 14/20 |
Each for $59.67
|
|
||||
|
31501152
|
Synthware™
F331425 |
25 mL | 14/20 |
Each for $60.97
|
|
||||
|
31501153
|
Synthware™
F331450 |
50 mL | 14/20 |
Each for $62.10
|
|
||||
|
31501154
|
Synthware™
F339100 |
100 mL | 19/22 |
Each for $68.30
|
|
||||
Description
- All sizes are hand blown from tubing to ensure uniform wall thickness
- Includes two hooks that are 180° apart
Specifications
| Short neck round bottom flask |