Learn More
Synthware™ Round Bottom Flasks with Hooks
Short neck with standard taper outer joint
Supplier:  Synthware™ F331410
 View more versions of this product
Catalog No. 31501151
Only null left
Explore available promotions
Description
Description
  • All sizes are hand blown from tubing to ensure uniform wall thickness
  • Includes two hooks that are 180° apart
Specifications
Specifications
Short neck round bottom flask
10 mL
14/20
Product Suggestions
Product Suggestions