Learn More
Synthware™ Rotary Evaporator Anti-Splash Adapters, Modified
GREENER_CHOICE
With two 4 mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask
$270.61 - $285.68
Specifications
Material Borosilicate Glass (3.3)
Product Type Anti-splash Adapter
View More Specs
Products 7
Catalog Number Mfr. No. Standard Taper Joint Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Capacity (Metric) Price Quantity & Availability  
31500630
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A602414
Upper: 24/40, Lower: 14/20 100 mL
Each for $270.61
Only null left
31500631
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A602419
Upper: 24/40, Lower: 19/22 100 mL
Each for $272.68
Only null left
31500632
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A602424
Upper: 24/40, Lower: 24/40 100 mL
Each for $273.96
Only null left
31500634
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A602924
Upper: 29/42, Lower: 24/40 100 mL
Each for $281.96
Only null left
31500635
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A601250
Upper: 24/40, Lower: 14/20 250 mL
Each for $272.25
Only null left
31500636
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A604250
Upper: 24/40, Lower: 24/40 250 mL
Each for $285.68
Only null left
31500633
GREENER_CHOICE
View Documents Promotion Details
Synthware™
A602914
Upper: 29/42, Lower: 14/20 100 mL
Each for $279.64
Only null left
Description
Description
Introduction: Anti-splash adapter is placed between the vapor tube and the evaporation flask on a rotary evaporator to prevent contents of the flask to be drawn into the condenser in the event of bumping
Specifications
Specifications
Borosilicate Glass (3.3)
Anti-splash Adapter