Learn More
Synthware™ Rotary Evaporator Anti-Splash Adapters, Modified
With two 4 mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask
Supplier: Synthware™ A601250
Description
Introduction: Anti-splash adapter is placed between the vapor tube and the evaporation flask on a rotary evaporator to prevent contents of the flask to be drawn into the condenser in the event of bumpingChemical Identifiers
| Borosilicate Glass (3.3) | |
| Anti-splash Adapter |
Specifications
| Upper: 24/40, Lower: 14/20 | |
| Borosilicate Glass (3.3) | |
| 250 mL | |
| Anti-splash Adapter |