Learn More
Synthware™ Claisen Adapters
Provides dual entry into a variety of flasks
$124.87 - $159.07
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass (3.3) |
| Product Type | Claisen Adapter |
| Quantity | 1 |
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity | |||||
|
31500681
|
Synthware™
A371420 |
14/20 |
Each for $124.87
|
|
|||||
|
31500682
|
Synthware™
A371922 |
19/22 |
Each for $127.37
|
|
|||||
|
31500683
|
Synthware™
A372440 |
24/40 |
Each for $144.50
|
|
|||||
|
31500684
|
Synthware™
A372942 |
29/42 |
Each for $159.07
|
|
|||||
Description
Synthware Claisen Adapter. Provides dual entry into a variety of flasks. Adapter has standard taper outer joints at top and on side and inner joint on bottom. All joints are the same size.Specifications
| Organic Synthesis | |
| Borosilicate Glass (3.3) | |
| Claisen Adapter | |
| 1 |