Learn More
Synthware™ Claisen Adapters
GREENER_CHOICE
Catalog No. 31500682
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
29/42
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31500682 19/22
31500681 14/20
31500683 24/40
31500684 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31500682 Supplier Synthware™ Supplier No. A371922
Only null left
Provides dual entry into a variety of flasks
Synthware Claisen Adapter. Provides dual entry into a variety of flasks. Adapter has standard taper outer joints at top and on side and inner joint on bottom. All joints are the same size.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Claisen Adapter
Quantity 1
Standard Taper Joint No. 19/22