Learn More
Synthware™ Claisen Adapters
Provides dual entry into a variety of flasks
Supplier: Synthware™ A372942
Description
Synthware Claisen Adapter. Provides dual entry into a variety of flasks. Adapter has standard taper outer joints at top and on side and inner joint on bottom. All joints are the same size.Specifications
| Organic Synthesis | |
| Claisen Adapter | |
| 29/42 |
| Borosilicate Glass (3.3) | |
| 1 |