Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and Glass Stopcock
Cylindrical style addition funnel with pressure equalizing arm and glass stopcock
$417.82 - $558.89
| Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Joint Number | Price | Quantity & Availability | ||||
31501713
![]() |
Synthware™
F621410G |
10 mL | 14/20 |
Each for $417.82
|
|
||||
31501715
![]() |
Synthware™
F621460G |
60 mL | 14/20 |
Each for $417.82
|
|
||||
31501714
![]() |
Synthware™
F621425G |
25 mL | 14/20 |
Each for $417.82
|
|
||||
31501716
![]() |
Synthware™
F622460G |
60 mL | 24/40 |
Each for $470.14
|
|
||||
31501717
![]() |
Synthware™
F621125G |
125 mL | 14/20 |
Each for $430.75
|
|
||||
31501718
![]() |
Synthware™
F624250G |
250 mL | 24/40 |
Each for $558.89
|
|
||||
Description
- Top outer and lower inner standard taper joints of same size
Specifications
| Pressure-equalizing Funnel with Glass Stopcock |