Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and Glass Stopcock
Cylindrical style addition funnel with pressure equalizing arm and glass stopcock
Supplier: Synthware™ F621425G
Description
- Top outer and lower inner standard taper joints of same size
Chemical Identifiers
| Pressure-equalizing Funnel with Glass Stopcock |
Specifications
| 25 mL | |
| 14/20 | |
| Pressure-equalizing Funnel with Glass Stopcock |