Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and Glass Stopcock
GREENER_CHOICE
Cylindrical style addition funnel with pressure equalizing arm and glass stopcock
Supplier:  Synthware™ F624250G
 View more versions of this product
Catalog No. 31501718
Only null left
Description
Description
  • Top outer and lower inner standard taper joints of same size
Specifications
Chemical Identifiers
Pressure-equalizing Funnel with Glass Stopcock
Specifications
250 mL
24/40
Pressure-equalizing Funnel with Glass Stopcock
Product Suggestions
Product Suggestions