Learn More
Synthware™ Inlet Adapters with Hose Connection
Hose connection for N2/Ar introduction or vacuum
$89.14 - $91.25
Specifications
Material Borosilicate Glass
Product Type Inlet Adapter with Hose Connection
View More Specs
Products 2
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
Catalog Number Mfr. No. Standard Taper Joint No. Price Quantity  
31500529
View Documents
Synthware™
A531420
14/20 N/A
Only null left
31500530
View Documents
Synthware™
A532440
24/40 N/A
Only null left
Description
Description
Supplied with one Nylon cap and one Viton O-ring
Specifications
Specifications
Borosilicate Glass
Inlet Adapter with Hose Connection
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'