Learn More
Synthware™ Inlet Adapters with Hose Connection
Hose connection for N2/Ar introduction or vacuum
Supplier:  Synthware™ A531420
 View more versions of this product
Catalog No. 31500529
Only null left
Explore available promotions
Description
Description
Supplied with one Nylon cap and one Viton O-ring
Specifications
Specifications
Borosilicate Glass
Inlet Adapter with Hose Connection
14/20
Product Suggestions
Product Suggestions